missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ Synergy™ H1 Hybrid Multi-Mode Monochromator Fluorescence Microplate Readers

Flexible monochromator-based detection with high performance dichroic-based filter detection. BioTek Synergy H1 Hybrid Multi-Mode Monochromator Fluorescence Microplate Readers with its monochromator optics supports top and bottom fluorescence intensity, UV-visible absorbance and high-performance luminescence detection.

Brand:  BioTek™ H1MD

Code : Z1

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 12538816

  • £28700.00 / Each


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Description

Description

  • Quadruple grating monochromator for maximum flexibility and ease of use
  • Compatible with Take3™ Multi-Volume Plate with 16 microspots of 2μL each for low-volume nucleic acid quantification
  • Dichroic-based filter optics for best performance and advanced detection technologies, such as fluorescence polarization and time-resolved fluorescence
  • Third generation quadruple grating design of the monochromator optics allows working at any excitation or emission wavelength with a 1nm step
  • Filter module includes its own light source and a high-performance dichroic-based wavelength selection system
  • Dual reagent injection system option automates ion channel assays and flash luminescent assays
  • Available with Gen5™ Data Analysis Software: reader control, advanced data analysis and flexible Excel export in one software package

Accessories

  • Take3 Multi-Volume Plate
  • BioStack™: 30 or 50 plate stacker
  • Gen5 Secure for 21CFR part 11 compliance
  • Fluorescence, Luminescence and Absorbance Test Plates

H1MFDG

  • Compatible with gas controller

Note

  • Dual reagent dispenser option available with all models
Applications
  • Academic Life Science Research
  • Biotech industry—especially smaller companies and start-ups
  • Clinical—Filter module for emerging applications of TRF, FP and luminescence
Specifications

Specifications

Multi-Mode Microplate Reader with Dispenser
2.0
39.1cm
22.5kg
Xenon flash
47.2cm
32.8cm
Monochromator fluorescence; Monochromator absorbance; Full-light luminescence; Dual reagent dispenser
Videos