missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ Synergy™ H1 Hybrid Multi-Mode Filter/Dichroic Fluorescence Microplate Readers

Configurable multi-mode microplate reader. BioTek′s patented Hybrid Technology offers high performance and applications versatility in a modular platform to expand as your laboratory′s needs change.

Brand:  BioTek™ H1F

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12153791

  • £19200.72 / Each


Estimated Shipment date
from Supplier 09-07-2020
Add to basket

Description

Description

  • Compatible with Take3™ Multi-Volume Plate with 16 microspots of 2μL each for low-volume nucleic acid quantification
  • Dichroic-based filter optics for best performance and advanced detection technologies, such as fluorescence polarization and time-resolved fluorescence
  • Filter module includes its own light source and high-performance dichroic-based wavelength selection system
  • Dual reagent injection system option automates ion channel assays and flash luminescent assays
  • Available with Gen5™ Data Analysis Software: reader control, advanced data analysis and flexible Excel export in one software package

Accessories

  • Take3 Multi-Volume Plate
  • BioStack™: 30 or 50 plate stacker
  • Gen5 Secure for 21CFR part 11 compliance
  • Fluorescence, Luminescence and Absorbance Test Plates

H1MFDG

  • Compatible with gas controller

Note

  • Dual reagent dispenser option available with all models
Applications
  • Academic Life Science Research
  • Biotech industry—especially smaller companies and start-ups
  • Clinical—Filter module for emerging applications of TRF, FP and luminescence
Specifications

Specifications

Filter/Dichroic Fluorescence Microplate Reader
Xenon flash
250 to 700nm
16nm
Filters: Fluorescein 0.25 pM
2.0
45°C
47.2cm
32.8cm
130w
Filter/dichroic fluorescence; Fluorescence polarization; Time-resolved fluorescence; Filtered luminescence
Gen5
CE and TUV marked, RoHS Compliant
H1F
0.2 at 5%
0.2 at 1%
1 to 384-well
PMT for monochromator and PMT for filter system
Sensitivity
±0.1%
5 Decades
5 to 1000μL
Linear, orbital, double orbital
39.1cm
22.5kg
130w
Yes
250 to 700nm
All the standard MicroPlate applications found in life science research laboratories
0 to 20%
1 to 19%
Videos
Documents