missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ Synergy™ H1 Hybrid Multi-Mode All Detection Modes Microplate Readers

Flexible monochromator-based detection with high performance dichroic-based filter detection. BioTek Synergy H1 Hybrid Multi-Mode All Detection Modes Microplate Readers have monochromator optics which supports top and bottom fluorescence intensity, UV-visible absorbance and high-performance luminescence detection.

Brand:  BioTek™ H1MFDG

Additional Details : Weight : 22.50000kg

 View more versions of this product

Product Code. 15366156

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Quadruple grating monochromator for maximum flexibility and ease of use
  • Compatible with Take3™ Multi-Volume Plate with 16 microspots of 2μL each for low-volume nucleic acid quantification
  • Dichroic-based filter optics for best performance and advanced detection technologies, such as fluorescence polarization and time-resolved fluorescence
  • Third generation quadruple grating design of the monochromator optics allows working at any excitation or emission wavelength with a 1nm step
  • Filter module includes its own light source and a high-performance dichroic-based wavelength selection system
  • Dual reagent injection system option automates ion channel assays and flash luminescent assays
  • Available with Gen5™ Data Analysis Software: reader control, advanced data analysis and flexible Excel export in one software package

Accessories

  • Take3 Multi-Volume Plate
  • BioStack™: 30 or 50 plate stacker
  • Gen5 Secure for 21CFR part 11 compliance
  • Fluorescence, Luminescence and Absorbance Test Plates

H1MFDG

  • Gas controller compatible
  • Gas module must be purchased separately (BT1210007 or BT12310008)

Note

  • Dual reagent dispenser option available with all models

Applications

  • Academic Life Science Research
  • Biotech industry—especially smaller companies and start-ups
  • Clinical—Filter module for emerging applications of TRF, FP and luminescence
Specifications

Specifications

Microplate Reader; Dispenser; Compatible w/ Gas Controller
47.2cm
Monochromator fluorescence, full-light luminescence, filter/dichroic, time resolved fluorescence, polarization, filtered luminescence; Dual reagent dispenser; Compatible w/Gas controller
Xenon flash
32.8cm
Videos