missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Supply Tubing Set for 50 TS Microplate Washer

Accessory for 50 TS microplate washer

Brand:  Agilent BioTek 4070510

Code : 03

Additional Details : Weight : 0.10000kg

Product Code. 10064592

  • £152.00 / Each
Estimated Shipment: 03-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • For non-buffer switching
  • Length: 1m (3.27 ft.)

Specifications

Specifications

Supply Tubing Set
50 TS Microplate Washer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers