missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BioTek™ Precision™ Microplate Pipetting System
Innovative replacement for manual pipetting
Brand: BioTek™ PRC384U
Code : Z1
Additional Details : Weight : 12.70000kg
Description
Features
- Available with multichannel manifold for continuous reagent dispensing
- Ideal for routine liquid deliveries such as reagent addition, plate replication, serial dilutions, sample transfer, and mixing within a well Configurable multistation platform and XY transport design provide effortless 96- to 384-well plate transfers
- Proprietary pipet technology provides reliable way to pick up and seal standard tips with controlled force Open design guarantees compatibility with robotic plate handlers
- Accommodates BioTek and other standard 50 to 250μL pipet tips; and 96- and 384-well flat-, round-, and V-bottom plates
Software
- Preinstalled software enables user to copy existing demonstration protocol and modify it; memory stores up to 80 program files User can create, edit, copy, and delete files using external keypad and 24-character LCD — virtually any liquid transfer parameter can be customized Configurable aspiration depth ensures that tips don't touch bottom of wells while performing sensitive cell assays
- Optional Precision Power™ graphical user interface (PRCPWRV2) simplifies program creation under Windows™ 98, NT, 2000, and XP
- Programs can be created and run from a PC, or downloaded and run from onboard microprocessor
Specifications
Microplate Pipetting System | |
≤1.5% CV typical at 100μL | |
100μL/well, 96-wells, 1 x 8 : 14 sec., 100μL/well, 96-wells, single-channel : 4 min. | |
12.7kg | |
38.1cm | |
100 to 240V | |
CE, TUV | |
≤0.7A | |
Manifold | |
Precision Power™, for PC protocol programming and execution |
96 and 384-well microplates | |
1% typical at 100μL | |
External membrane keypad with 25 alphanumeric keys, two-line x 24-character LCD | |
40.64cm | |
50/60Hz | |
5 to 120μL | |
100 to 240V 50/60Hz | |
80% | |
15° to 40°C | |
2L |