missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ Precision™ Microplate Pipetting System

Innovative replacement for manual pipetting

Brand:  BioTek™ PRC384M

Code : Z1

Additional Details : Weight : 12.70000kg

 View more versions of this product

Product Code. 11503565

  • £18000.00 / Each


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Specifications

Specifications

Microplate Pipetting System
≤1.5% CV typical at 100μL
100μL/well, 96-wells, 1 x 8 : 14 sec., 100μL/well, 96-wells, single-channel : 4 min.
12.7kg
38.1cm
100 to 240V
CE, TUV
≤0.7A
Manifold
Precision Power™, for PC protocol programming and execution
96 and 384-well microplates
1% typical at 100μL
External membrane keypad with 25 alphanumeric keys, two-line x 24-character LCD
40.64cm
50/60Hz
5 to 120μL
100 to 240V 50/60Hz
80%
15° to 40°C
2L
Videos
View accessory products