missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ PowerWave™ Microplate Spectrophotometer

Designed to run 24 hours a day, 7 days a week for use as a stand-alone instrument or in an automated system. BioTek PowerWave Microplate Spectrophotometers are suitable for any UV/Vis application between 200 to 999nm including direct DNA/RNA quantitation and DNA purity.

Brand:  BioTek™ RPRWI

Code : 04

Additional Details : Weight : 50.00000kg

Product Code. 10340763

  • £17901.97 / Each


Estimated Shipment date
from Supplier 09-07-2020
Add to basket

Includes: Gen 5 data analysis software, serial cable (null modem, 9- to 95-pin female), bidirectional RS-232 interface

Description

Description

Key Features

  • Feature monochromator with xenon flash lamp, 5nm bandpass, user-programmable shake timing and frequency and 4-Zone™ natural convection incubation to 50°C
  • Quickly and accurately read 96- or 384-well microplates, depending on model
  • With PowerWave and Gen5™ software, use automatic pathlength correction for direct nucleic acid quantification or to convert cuvet-based assays into microplate format
  • Also accepts 1cm Bio-Cell™ vessels for standard spectrophotometer uses when used with optional adapter (BT-7270512)

Additional Features

  • Select wavelengths in 1nm increments—without the use of filters
  • Read endpoint, kinetic and spectral scans, and well scans in three read modes (normal, rapid and sweep)
  • Quickly perform spectral scans and analysis of multiple wells with overlay scans for fast comparisons
  • Allow automatic pathlength correction to 1cm values for high-throughput nucleic acid analysis
  • Compatible with automated systems
  • Conforms to regulatory requirements for in vitro diagnostic use
  • Electrical requirements: 100–240VAC 50/60Hz

Software Features
  • Powerful Gen5™ control and analysis software automatically runs the reader, analyzes the results and generates a customized report, all with one mouse click
  • Choose optional Gen5 Secure software when compliance to 21CFR Part 11 is essential

Certifications

CE marked; ETL listed

Specifications

Specifications

Microplate Spectrophotometer
<1% ± 0.10 OD typical
Gen 5 data analysis software, serial cable (null modem, 9- to 95-pin female), bidirectional RS-232 interface
±1% typical
Programmable shaking
Videos
Documents
View accessory products