Learn More
Agilent BioTek MultiFlo FX Microplate Dispenser Modules
Automated reagent dispenser for 6- to 1536-well microplates.
£5085.00 - £8895.00
Specifications
For Use With (Equipment) | MultiFlo FX Microplate Dispenser |
---|---|
Quantity | 1/Cs. |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
12578816
|
Agilent BioTek
7210010 |
Peristaltic Pump Dispenser Module |
£5085.00
Each |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12588816
|
Agilent BioTek
7210008 |
Dual Syringe Drive Dispenser (Auto) |
£8895.00
Each |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12598816
|
Agilent BioTek
7210009 |
Dual Syringe Drive Dispenser (Non-Auto) |
£6360.00
Each |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Automated reagent dispenser for 6- to 1536-well microplates
- Parallel Dispense technology allows up to four independent reagents to be dispensed in parallel without potential carryover
- RAD technology for dispensing to individual wells
- Cell friendly angled dispense and/or wash tubes won't disrupt cell layers
Applications:
- For benchtop and integrated automation workflows
- Bulk reagent dispensing
- Cell seeding
- Cell washing /media exchange
- Bead washing
Specifications
MultiFlo FX Microplate Dispenser | |
1/Cs. |