missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ MultiFlo™ FX Microplate Dispensers

Modular and automated for 6- to 1536-well plates offering Parallel Dispense™ technology

Brand:  BioTek™ MFXW

Additional Details : Weight : 6.50000kg

 View more versions of this product

Product Code. 15376196

  • £12100.00 / Each


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Description

Description

MultiFlo FX is a fully modular, automated reagent dispenser for 6- to 1536-well plates offering BioTek's unique Parallel Dispense technology - both peristaltic and syringe pump operation down to 500nL. An available microplate wash module adds multi-purpose functionality. A fully configured MultiFlo FX replaces up to five liquid handlers.

  • Four independent reagents can be dispensed in parallel without potential carryover
  • Parallel Dispense technologies: Choice of peristaltic or syringe pump
  • Onboard computer with two USB flash drive ports for file transfer, storage and operation
  • Small footprint allows the usage in biosafety cabinet or integrated into an automated robotic system
  • Programmable functions and values displayed via touch screen user interface
  • Compatible with wide range of plate formats up to 50mm in height
  • Operating Temperature: 15° to 35°C (59° to 95°F)
  • Dimensions: D x W x H: 10.6 x 16.1 x 7.6 in. (26.9 x 40.9 x 19.3cm)

MFX:

  • Base unit, unconfigured
MFXP:
  • With one peri-pump dispenser
  • Manifold Types: 8-tip cassette (1 x 8)
  • Volume Range: 500nL to 3,000μL/well
  • Flow Rates: Low, medium or high
  • Cassette Size: 1, 5 and 10μL
MFXW:
  • With wash module (include 96/384 wash manifold)
  • Manifold Types: 96-well washing (8-tube (1x8) manifold; 12-tube (1x12) manifold; 96-/384-well washing (Dual-Action 16-tube (1x16) manifold)
  • Volume Range: 20-30,000μL/well
  • Flow Rates: 1 to 9
  • Wash Cycles: 1 to 10
MFXPW:
  • One peri-pump dispenser
  • Wash module

Specifications

Specifications

50/60Hz
CE, TUV
3%
10μL/well, 384-wells: 8 sec., 10μL/well, 96-wells: 3 sec., 5μL/well, 384-wells: 6.5 sec., 1μL/ well, 1536-wells: 21 sec.
For 6 to 1536-well plates
80%
5.7 in. LCD touchscreen display
15° to 35°C
Liquid Handling Control™
3 aspiration/disp. cycles: >300μL/well, 96-wells, 8-tube manifold, final aspirate <105 sec.
40.89cm
100 to 240VAC
26.92cm
<3% CV (96-/384-well plates; 300μL/well), <3% CV (12-/48-well plates; volume dependent), <5% CV (6-well plates; 5560μL/well)
100 to 240VAC 50/60Hz
19.3cm
Two USB flash drive ports, wash module
96-well washing (8-tube (1 x 8) manifold, 12-tube (1 x 12) manifold, 96/384-well washing (dual-action 16-tube (1 x 16) manifold)
<2μL/well, 300μL dispense, 0.1% Tween
2L
8.8kg
With wash module
Videos