missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes

Dispense Cassettes for use with MultiFlo FX Dispenser and EL406 Washer Dispenser.

Brand:  Agilent BioTek 7170016

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 12551547

  • £1510.00 / Each
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Volume range 1 μL to 10 mL
  • Accommodates 6- to 1536-well microplates, PCR trays, deep-well microplates, microtubes and other tube configurations up to 4 in. in height
  • Dispense 1536 wells in 18 s.
  • Cassettes can be autoclaved without recalibration
  • Dispense up to eight reagents at one time
  • Customizable dispense pattern allows skipped columns or rows
  • Built-in priming trough
  • Easy maintenance of individually replaceable tips and choice of replacement cassettes or tubing
  • Compatible with Bio-Stack microplate stacker 11-120-601 for increased throughput

Specifications

Specifications

0.0075 in.
For liquid handling
0.019 cm
Stainless Steel
1 to 50 μL
Low Volume 1536 Well Cassette
1 μL
EL406 Microplate Washer, Multiflo Dual Syringe Dispenser Module, Agilent BioTek MultiFlo FX Multimode Dispenser
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers