missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ Gas Controllers Ready Synergy™ H1 Hybrid Multi-Mode Microplate Reader

Flexible monochromator-based multi-mode microplate reader that can be turned into a high-performance Hybrid system with the addition of a filter-based optical module.

Brand:  BioTek™ H1FG

Additional Details : Weight : 22.50000kg

 View more versions of this product

Product Code. 15346156

  • £18590.00 / Each


Estimated Shipment date
from Supplier 25-04-2019
Add to basket

Description

Description

  • Monochromator optics uses third generation quadruple grating design which allows working at any excitation or emission wavelength with a 1nm step
  • Supports top and bottom fluorescence intensity, UV-visible absorbance and high performance luminescence detection
  • Completely independent filter module; includes its own light source, and a high performance dichroic-based wavelength selection system
  • Supports advanced detection modes such as Fluorescence Polarization, Time-Resolved Fluorescence & TR-FRET and filtered luminescence (e.g. BRET)
  • Compatible with the Gas Controller for control and monitoring of CO2 and O2
  • Compatible with Take3™ Micro-Volume Plate: samples down to 2μL volume can be measured; especially useful when working with precious samples, for fast and accurate DNA/RNA quantification at 260nm
  • Includes Gen5 software for reader control, advanced data analysis and flexible Excel export in one software package
Specifications
  • Read methods: Endpoint, kinetic, spectral scanning, well area scanning
  • Microplate types: 6- to 384-well plates
  • Shaking; Linear, orbital, double orbital
  • Automation: BioStack and 3rd party automation compatible; BioSpa 8 Automated Incubator compatible
  • CO2 and O2 control Range: 0 to 20% (CO2); 1 to 19% (O2)
  • CO2 and O2 control Resolution: +0.1% (CO2 and O2)
  • CO2 and O2 control Stability: +0.2% at 5% CO2; +0.2% at 1% O2
  • Power: 130w max.
  • Dimensions (D x W x H): 47.2 x 39.1 x 32.8cm)
  • Weight: 22.5kg
Model: BTH1FG
With Full-light luminescence, Filter/dichroic fluorescence, Fluorescence polarization, Time resolved fluorescence, Filtered luminescence, Temperature control to 45°C
 
Model: BTH1FDG
With Full-light luminescence, Filter/dichroic fluorescence, Fluorescence polarization, Time resolved fluorescence, Filtered luminescence, Temperature control to 45°C
Dual reagent injection system to automate inject/read assays such as ion channels assays or flash luminescence assays
 
Model: BTH1FMG
With Monochromator fluorescence, Monochromator absorbance, Full-light luminescence, Filter/dichroic fluorescence, Fluorescence polarization, Time resolved fluorescence, Filtered luminescence, Temperature control to 45°C

Order Info

Gas module must be purchased separately (BT1210007 or BT12310008)

Certifications

CE and TUV marked

Compliance

RoHS Compliant

Specifications

Specifications

Multi-Mode Microplate Reader
6- to 384-well plates
Photodiode, PMT
200 to 700nm fluorescence intensity, 300 to 700nm luminescence, 320 to 700nm fluorescence polarization, 200 to 700nm time resolved fluorescence
2.5 pM to 4 pM monochromators, 0.25 pM filters fluorescence, 20 amol ATP monochromator, 10 amol ATP filter luminescence, 1.2 mP fluorescence polarization, Europium 40 fM with filters, 1200 fM with monos time resolved fluorescence
45°C
96 wells: 11 seconds; 384 wells: 22 seconds
39.1cm
22.5kg
RoHS Compliant
Gen5™ Data analysis
H1FG
Proprietary Hybrid technology
Xenon flash
High performance dichroic-based
Full-light luminescence, Filter/dichroic fluorescence, Fluorescence polarization, Time resolved fluorescence, Filtered luminescence, Temperature control to 45°C
0 to 4 O.D.
Linear, orbital, double orbital
47.2cm
32.8cm
130w max.
Gas Controllers
CE and TUV marked
Videos