Learn More
BioTek™ FLx800™ Microplate Fluorescence Readers
Choose from models with top probe or top and bottom probes.
Brand:  BioTek™ FLX800TB
Code : 04
Additional Details : Weight : 13.60000kg
 View more versions of this product
Product Code. 11583515
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Includes: Probe(s), 485 and 360nm excitation filters, 528 and 460nm emission filters, serial and power cables, power cord
Description
Description
  • All models read 6-, 12-, 24-, 48-, 96-, and 384-well plates
  • Onboard software stores 55 programmable protocols, up to 75 custom programs, and eight microplate assay results
  • Two-line, 24-character LCD and menu-driven software simplify operation and programming
  • Onboard diagnostic testing utilizes three methods — an optical self-test, a universal test plate, and a “wet test” — to check instrument performance and optical specifications, and provides either soft or hardcopy results
  • Fiber-optic probes provide excellent sensitivity over a wide range
  • All units feature convenient front-panel access to accessories
Software Features
  • Includes Gen5RC reader control software or Gen5 Data Analysis software (model dependent)
  • Onboard data analysis provides curve-fitting options, including linear regression, polynomial, point-to-point, cubic spline, and four-parameter logistic curve fits
Model Features
  • Top probe is ideal for reading solid black plates
  • Bottom probe reads covered plates
  • Combination of top and bottom probes offers the most flexibility and includes automated probe switching for added convenience
  • Extended-range photomultiplier tube (PMT) extends emission range to 800nm Shaking and incubation options are ideal for temperature-sensitive assays
  • Single reagent injector is suitable for ion-uptake assays and flash luminescence
Note
Supplied filters can be exchanged for other filters at time of purchase at no extra cost so that you can configure the unit to meet your specific application
Order Info
Optional Gen5 software automatically runs the reader, analyzes the results, and generates a customized report. Or choose Gen5 Secure software to meet requirements of 21 CFR Part 11.
Certifications
CE marked and ETL listed, Certified to CAN/CSA C22.2 No. 1010.1-92 and EN61010-1, including amendments 1 and 2
Compliance
Conforms to UL 3101-1
Specifications
Specifications
Quartz halogen, 1000-hour life
300 to 650nm
340 to 650nm
5 decades (0 to 99,999)
23cm
100-240VAC; 24V external transformer
Top and bottom probes, 485 and 360nm excitation filters, 528 and 460nm emission filters, serial and power cables, power cord
Fluorescence, Luminescence
485 and 360 excitation filters
528 and 460 emission filters
39cm
50/60Hz
100-240VAC; 24V external transformer 50/60Hz
Gen5RC reader control software
Videos
Documentation
Technical Documents
View accessory products