BioTek™ FLx800™ Microplate Fluorescence Readers

Choose from models with top probe or top and bottom probes.

Overview
Brand: BioTek™

Manufacturer Part Number: FLX800T

FLx800T Top Filter Fluorescence, Luminescence andFilter Luminescence, Gen5 software included.

UNSPSC: 41113037

Code: 04

Additional Details:
Additional Details: Weight: 13.60000kg



Includes: Choose from models with top probe or top and bottom probes.

Product Code. 11573515

Quantity Price
1 £ 12200.0 / Each
Estimated Shipment date
from Supplier 28-03-2017
Add to basket


  • Description and Specification
  • Documentation
  • Accessories and Spare Parts
Description and Specification

Specification

Excitation Wavelength Range 300 to 650nm
Detector Type Fluorescence, Luminescence
Emission Wavelength Range 340 to 650nm
Hertz 50/60Hz
Emission Filters 528 and 460 emission filters
Width (Metric) 39cm
Software Type Gen5RC reader control software
Light Source Quartz halogen, 1000-hour life
Excitation Filters 485 and 360 excitation filters
Dynamic Range 5 decades (0 to 99,999)
Height (Metric) 23cm
Voltage 100-240VAC; 24V external transformer
Electrical Requirements 100-240VAC; 24V external transformer 50/60Hz
Includes Top probe, 485 and 360nm excitation filters, 528 and 460nm emission filters, serial and power cables, power cord

  • All models read 6-, 12-, 24-, 48-, 96-, and 384-well plates
  • Onboard software stores 55 programmable protocols, up to 75 custom programs, and eight microplate assay results
  • Two-line, 24-character LCD and menu-driven software simplify operation and programming
  • Onboard diagnostic testing utilizes three methods — an optical self-test, a universal test plate, and a “wet test” — to check instrument performance and optical specifications, and provides either soft or hardcopy results
  • Fiber-optic probes provide excellent sensitivity over a wide range
  • All units feature convenient front-panel access to accessories

Software Features

  • Includes Gen5RC reader control software or Gen5 Data Analysis software (model dependent)
  • Onboard data analysis provides curve-fitting options, including linear regression, polynomial, point-to-point, cubic spline, and four-parameter logistic curve fits

Model Features

  • Top probe is ideal for reading solid black plates
  • Bottom probe reads covered plates
  • Combination of top and bottom probes offers the most flexibility and includes automated probe switching for added convenience
  • Extended-range photomultiplier tube (PMT) extends emission range to 800nm Shaking and incubation options are ideal for temperature-sensitive assays
  • Single reagent injector is suitable for ion-uptake assays and flash luminescence

Note
Supplied filters can be exchanged for other filters at time of purchase at no extra cost so that you can configure the unit to meet your specific application

Order Info: Choose from models with top probe or top and bottom probes.

Compliance: Choose from models with top probe or top and bottom probes.

Certifications: Choose from models with top probe or top and bottom probes.

Documentation

Technical documents

Accessories and Spare Parts