missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Filters for ELx808 Ultra Microplate Reader
Accessory for use with ELx808 Ultra Microplate Readers.
£750.00
Specifications
Product Type | Filter |
---|---|
For Use With (Equipment) | ELx808 Ultra Microplate Readers |
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
11523595
|
Agilent BioTek
3404750 |
750 nm |
£750.00
Each |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10116562
|
Agilent BioTek
3100450 |
450 nm |
£750.00
Each |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Accessory for use with ELx808 Ultra Microplate Readers.Specifications
Filter | |
ELx808 Ultra Microplate Readers |