Learn More
Agilent BioTek Filters for ELx800 Universal Microplate Reader
Accessory for use with ELx800 Universal Microplate Readers.
Brand: Agilent BioTek 7334570
Code : 04
Additional Details : Weight : 0.10000kg
Description
Accessory for use with ELx800 Universal Microplate Readers.Specifications
Filter | |
ELx800 Universal Microplate Readers | |
570 nm |