Learn More
BioTek™ ELx808™ Absorbance Microplate Readers
Store up to 55 user-programmable protocols. BioTek ELx808 Absorbance Microplate Readers read 96-well microplates and provide precise, accurate, and repeatable results.
Brand:  BioTek™ ELX808IU
Code : 03
Additional Details : Weight : 13.60000kg
 View more versions of this product
Product Code. 10665053
  • £6206.20 / Each
Estimated Shipment date
from Supplier 10-07-2018
Add to basket
Includes: Filters, tungsten halogen light source, parallel printer cable, null modem serial cable and Gen5RC reader control software
Description
Description
  • Readers can perform multiple assays per plate
  • On-board diagnostic self-test may be used with an optional calibration test plate to easily confirm and document performance
  • Kinetic rate, R2, or onset time is reported without a PC
  • Linear well scanning is ideal for agglutination assays
  • Staggered optical design eliminates channel-to-channel crosstalk
  • Reference channel rolls optical signal for uniform channel-to-channel signal
  • Curve-fitting options include linear, 4-P, 2-P (logit/log), cubic, quadratic, cubic spline, and point-to-point
  • Measurements can be read in intervals as short as six seconds
  • Additional data analysis options include cutoff formulas, endpoint, and kinetic analysis Supplied with 4-Zone™ Temperature Control
  • Extended dynamic range
Model ELx808
  • Data analysis options include data transformation formulas, assay validation, and control validation
  • Optional 4-Zone™ Temperature Control improves reproducibility in enzyme kinetic reactions
Order Info
Optional Gen5 Data Analysis Software (Mfr. No. GEN5) provides simple, fast data collection, calculation, exporting, and reporting. Or select Gen5 Secure software to meet requirements of 21 CFR Part 11.
Certifications
CE marked
Specifications
Specifications
340 to 900nm
>lt;1% at 2.5 O.D., >lt;2% at 3.5 O.D.
39.4cm
13.6kg
>lt;1% at 2.5 O.D. (typical)
96 wells: 12 seconds (single wavelength), 20 seconds (dual wavelength), or 6 seconds (kinetic single wavelength)
22.2cm
Filters, tungsten halogen light source, parallel printer cable, null modem serial cable and Gen5RC reader control software
Videos
Documentation
Technical Documents