missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ ELx800™ Absorbance Microplate Readers

Store up to 75 custom assay definitions and eight test results.

Brand:  BioTek™ ElX800UV

Code : 03

Additional Details : Weight : 20.00000kg

 View more versions of this product

Product Code. 10320053

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Includes: Gen5RC reader control software

Description

Description

  • Two 24-character LCD and convenient soft-keys for easy use
  • Onboard data analysis offers extensive curve-fitting options: linear, cubic, quadratic, 4-P, log-logit, cubic-spline, and point-to-point; plus endpoint and kinetic measurement
  • Three modes of performance and optical specifications testing: optical self-test, a calibration test plate, and a “wet test”
  • Auto or manual plate layout for blanks, controls, standards, and samples
  • Compatible with optional Gen5™ software
  • USB and RS232 ports for PC connection; parallel printer port
Features
  • ELx800 has 24-, 48-, and 96-well microplate read modes
  • ELx800UV contains optics to extend wavelength range down to 340nm
  • ELx800NB has the ability to read 384-well and 60-/72-/96-well Terasaki plates (with the help of optional software, sold separately)

Order Info

Optional Gen5 Data Analysis Software (Mfr. No. GEN5) provides simple, fast data collection, calculation, exporting, and reporting. Or choose Gen5 Secure software to meet requirements for 21 CFR Part 11 (Mfr. No. GEN5SECURE). 100-240VAC 50/60Hz.

Certifications

CE listed; ETL listed

Specifications

Specifications

340 to 750nm
±1%, ±0.010Abs (from 0 to 2.000Abs @405nm)
50/60Hz
100-240V 50/60Hz
±1% (from 0 to 2.000Abs @405nm), ±3% (from 2.000 to 3.000Abs @405nm)
96 wells: 30 seconds
100-240V
Videos
Documentation

Technical Documents