Learn More
BioTek™ ELx50™ Microplate Strip Washer
Syringe-drive fluid delivery system for precise control
Brand:  BioTek™ ELX5012
Code : 03
Additional Details : Weight : 9.80000kg
 View more versions of this product
Product Code. 10524513
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Includes: QC report and performance verification data sheet; Aerosol cover; Built-in pumps; Accessory package including bottles, tubing, and maintenance kit
Description
Description
Key Features
  • Washes single strips or full plates
  • Efficient washing ensures low background absorbance and small residue volumes
  • Self-contained and programmable
  • Micro-controlled, syringe-drive positive-displacement pump fluid delivery system for precise control
  • Biomagnetic separation and vacuum filtration options available
  • Safety features include aerosol cover, built-in decontamination routine, and isolation of electronics from fluidics
  • Biomagnetic separation and vacuum filtration options available
  • Choice of 8-, 12- or 16-channel manifold models
  • Optional switching of up to three wash buffers
Applications:
Suitable for EIA, fluorescence, chemiluminescence, RIA, DNA probe, and cellular assays
Certifications
CE listed; ETL listed
Specifications
Specifications
1 to 10
LCD
35cm
16.25cm
QC report and performance verification data sheet; Aerosol cover; Built-in pumps; Accessory package including bottles, tubing, and maintenance kit
Standard Strip Washer
25 to 3000μL/well
CE and ETL marked
150 to 1000μL/well/sec.
100 to 250VAC 50/60Hz
1 to 600 sec.
RS-232
40cm
8.9kg
50/60Hz
100 to 250VAC
<4μL/well
Suitable for EIA, fluorescence, chemiluminescence, RIA, DNA probe, and cellular assays
12
Videos
Documentation
Technical Documents
For in vitro diagnostic use