BioTek™ ELx50™ Microplate Strip Washer

Syringe-drive fluid delivery system for precise control

Overview
Brand: BioTek™

Manufacturer Part Number: ELX5012V

UNSPSC: 41103207

Code: 03

Additional Details:
Additional Details: Weight: 9.80000kg



Disclaimers: For in vitro diagnostic use

Product Code. 10034212

Quantity Price
1 £ 5100.0 / Each
Estimated Shipment date
from Supplier 03-04-2017
Add to basket


Description and Specification

Specification

Flow Rate 150 to 1000μL/well/sec.
Soak Time 1 to 600 sec.
Automation Interface RS-232
Volume (Metric) Dispense 25 to 3000μL/well
Depth (Metric) 40cm
Hertz 50/60Hz
Width (Metric) 35cm
Height (Metric) 16.25cm
Type Washer with valves and automatic buffer switching
Display LCD
Weight (Metric) 8.9kg
Volume (Metric) Residual <4μL/well
Includes QC report and performance verification data sheet; Aerosol cover; Built-in pumps; Accessory package including bottles, tubing, and maintenance kit
Well Count 12
Voltage 100 to 250VAC
Wash Cycles 1 to 10
For Use With (Application) Suitable for EIA, fluorescence, chemiluminescence, RIA, DNA probe, and cellular assays
Electrical Requirements 100 to 250VAC 50/60Hz
Certifications/Compliance CE and ETL marked
Documentation

Technical documents