missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ ELx405™ Select Deep-Well Microplate Washer

World's first deep well microplate washer that aspirates and dispenses into 96- and 384-well plates <50mm in height

Brand:  BioTek™ ELX405UCWD

Additional Details : Weight : 13.50000kg

 View more versions of this product

Product Code. 12558846

  • £16400.00 / Each


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Installed on a lab bench or integrated within a robotic system, the ELx405 Select lowers background values and increases important signal-to-noise ratios.

  • Accommodates 96- and 384-deep well microplates as well as ANSI/SBS standard and deep-well plates ≤50mm
  • Polystyrene and magnetic bead assays along with standard ELISAs
  • Patented Dual-Action™ manifold provides independent control of the aspirate and dispense manifolds for precise overfill washing and overflow protection
  • Internal positive displacement pump eliminates the use of troublesome pressure delivery systems and allows the use of any size dispense reservoir
  • Built-in priming trough allows automatic manifold priming and rinsing during both operation and maintenance modes and eliminates the need for blank plates when used in automated robotic systems
  • Fully variable dispense volumes, flow, and aspiration rates provide total control of the washing cycles from a gentle cell-washing application to vigorous wash for high-binding assays
  • Built-in fluid flow, fluid detection, and vacuum sensing provide complete protection for unattended operation
  • BioStack™ compatible for automated washing of up to 50 ANSI/SBS standard height microplates
  • Compatible with Liquid Handling Control Software for additional flexibility

Applications:

ELISAs, MSD assays, HCS immune cytochemistry, FLIPR Ca+ flux, Cell-based assays, Magnetic and polystyrene bead assays, Gene expression assays, Cytokine assays, ELISPOT assays, Plasmid DNA purification, Serum/plasma sample preparation, Cell signaling – phospho flow setup for flow cytometry

Specifications

Specifications

Built-in ultrasonic bath for protein and salt crystal buildup; programmable up to 60 minutes (model dependent)
Programmable in minutes and seconds, up to 60 min.
35.6cm
25.4cm
Dual-Action 96-tube manifold and optimized cell washing
100 to 240V
CE and ETL marked
<8A
10 to 80%, noncondensing
100 to 240V 50/60Hz
≤3% CV
Backlit LCD
43.2cm
13.5kg
50/60Hz
≤2μL/well
In Vitro diagnostic use
One positive displacement pump
59° to 86°F
Videos

For in vitro diagnostic use