Learn More
BioTek™ ELx405™ Select Deep-Well Microplate Washer
World's first deep well microplate washer that aspirates and dispenses into 96- and 384-well plates <50mm in height
Brand: BioTek™ ELX405UCWSD
Code : Z1
Additional Details : Weight : 13.50000kg
Description
Installed on a lab bench or integrated within a robotic system, the ELx405 Select lowers background values and increases important signal-to-noise ratios.
- Accommodates 96- and 384-deep well microplates as well as ANSI/SBS standard and deep-well plates ≤50mm
- Polystyrene and magnetic bead assays along with standard ELISAs
- Patented Dual-Action™ manifold provides independent control of the aspirate and dispense manifolds for precise overfill washing and overflow protection
- Internal positive displacement pump eliminates the use of troublesome pressure delivery systems and allows the use of any size dispense reservoir
- Built-in priming trough allows automatic manifold priming and rinsing during both operation and maintenance modes and eliminates the need for blank plates when used in automated robotic systems
- Fully variable dispense volumes, flow, and aspiration rates provide total control of the washing cycles from a gentle cell-washing application to vigorous wash for high-binding assays
- Built-in fluid flow, fluid detection, and vacuum sensing provide complete protection for unattended operation
- BioStack™ compatible for automated washing of up to 50 ANSI/SBS standard height microplates
- Compatible with Liquid Handling Control Software for additional flexibility
Applications:
ELISAs, MSD assays, HCS immune cytochemistry, FLIPR Ca+ flux, Cell-based assays, Magnetic and polystyrene bead assays, Gene expression assays, Cytokine assays, ELISPOT assays, Plasmid DNA purification, Serum/plasma sample preparation, Cell signaling – phospho flow setup for flow cytometry
Certifications
CE and ETL marked
Specifications
Built-in ultrasonic bath for protein and salt crystal buildup; programmable up to 60 minutes (model dependent) | |
Programmable in minutes and seconds, up to 60 min. | |
35.6cm | |
25.4cm | |
Dual-Action 96-tube manifold, optimized cell washing and built-in ultrasonic cleaning | |
100 to 240V | |
CE and ETL marked | |
<8A | |
10 to 80%, noncondensing | |
100 to 240V 50/60Hz |
≤3% CV | |
Backlit LCD | |
43.2cm | |
13.5kg | |
50/60Hz | |
≤2μL/well | |
In Vitro diagnostic use | |
One positive displacement pump | |
59° to 86°F |
For in vitro diagnostic use