Learn More
BioTek™ EL406 Microplate Washer/Dispenser
Fast, full plate washing capability using patented Dual Action™ manifold
Brand:  BioTek™ 406PSUB2SB
Additional Details : Weight : 14.50000kg
 View more versions of this product
Product Code. 15306186
  • £47500.00 / Each
Estimated Shipment date
from Supplier 29-05-2018
Add to basket
Description
Description
  • Fast, full microplate washing, optimized for loosely adherent cell monolayers
  • Up to three reagent dispensers with choice of either peristaltic pump or syringe drive technologies for different assay requirements 96-, 384- and 1536-well microplates
  • Automates long and tedious assay liquid-handling processes
  • Smaller benchtop or robotic system footprint compared to separate washer and three dispensers
  • BioStack™ compatible for automated processing of up to 50 standard height microplates
  • Full control with powerful Liquid Handling Control™ PC Software
  • Optimized washing for loosely adherent cell layers
  • Built-in, patent-pending Ultrasonic Advantage™ for unattended wash manifold maintenance
  • Optional buffer switching of up to four buffers for complex wash routines
Applications:
  • ELISA automation
  • MSD automation
  • High-content screening immunocytochemistry
  • Label-free cell-based assays — especially 1536-well density calcium flux assays
  • Magnetic and polystyrene bead assay
  • Automation for gene and protein expression
Specifications
Specifications
384-, 1536-well (small bore sapphire jeweled tubes)
Programmable in minutes and seconds, up to 60 min.
42cm
31.8cm
50/60Hz
100 to 240V
< 2μL/well
Fluorescence intensity, time-resolved fluorescence, fluorescence polarization, luminescence, UV-visible absorbance, FRET, TR-FRET, BRET, well area
≤3% CV
13 seconds, 300μL/well, 96-wells; 17 seconds, 100μL/well, 384-wells (192-tube manifold)
46cm
Wash Buffer switching, 384- and 1536-tube manifolds; Small bore, sapphire jeweled tubes
384- and 1536-tube manifolds
25 to 3000μL/well
CE and ETL marked
100 to 240V 50/60Hz
Videos