missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ EL406 Microplate Washer/Dispenser

Fast, full plate washing capability using patented Dual Action™ manifold

Brand:  BioTek™ 406PSUB1

Code : Z1

Additional Details : Weight : 14.50000kg

 View more versions of this product

Product Code. 11573595

  • £37000.00 / Each


Estimated Shipment date
from Supplier 22-02-2018
Add to basket

Specifications

Specifications

96-well
Programmable in minutes and seconds, up to 60 min.
42cm
31.8cm
50/60Hz
100 to 240V
< 2μL/well
Fluorescence intensity, time-resolved fluorescence, fluorescence polarization, luminescence, UV-visible absorbance, FRET, TR-FRET, BRET, well area
≤3% CV
13 seconds, 300μL/well, 96-wells; 17 seconds, 100μL/well, 384-wells (192-tube manifold)
46cm
Wash buffer switching; Three reagent dispensers; 96-tube manifold
96-tube manifold; Dual Action manifold
25 to 3000μL/well
CE and ETL marked
100 to 240V 50/60Hz
Videos