Learn More
BioTek™ EL406 Microplate Washer Dispenser
Offers fast, full plate washing along with three reagent dispensers in one, compact instrument
Brand:  BioTek™ 406PSNA4
Additional Details : Weight : 14.50000kg
 View more versions of this product
Product Code. 15336176
  • £27500.00 / Each
Estimated Shipment date
from Supplier 26-06-2018
Add to basket
Description
Description
  • For 96-well microplates
  • Magnetic and polysyrene microsphere assays
  • ELISAs and cell-based assays
  • High-energy neodymium iron boron magnets
  • Selection of optimized field gradients within wells
  • Rapid separation of micrometer and nanometer beads with superior retention
  • Fast and efficient vacuum filtration
  • Fast microplate washing and dispensing
  • Patented Dual-Action™ wash manifold
  • Up to three reagent dispensers
  • Unattended operation for long and tedious liquid handling processes
  • Smaller benchtop or robotic system footprint compared to separate washer and three dispensers
  • BioStack™ compatible for automated processing of up to 50 standard height microplates
  • Choice of software control from instrument keypad or Liquid Handling Control™ (LHC) PC software
  • Built-in ultrasonic cleaner
  • Automatic switching of up to four wash buffers or reagents
Specifications
Specifications
96-well Microplates
406PSNA4
Videos