Learn More
Agilent BioTek Angled Tip Injector Head for Imaging Multi-Mode Reader
For use with Imaging Multi-Mode Readers.
Brand: Agilent BioTek 1320514
Additional Details : Weight : 1.00000kg
Description
- 21° Angled tips for dispensing toward sides of microplate well to prevent cell layer disruption
- Includes installation instructions
Specifications
Angled Tip Injector Head | |
Imaging Multi-Mode Reader | |
Dispensing towards sides of microplate well to prevent cell layer disruption. |