BioTek™ Cytation 5™ Multi-Mode Reader

For use with 6- to 384-well microplates. BioTek Cytation 5 Multi-Mode Readers work on Patented Hybrid Technology™ in multi-mode detection which combines high performance filters with variable bandwidth monochromators for versatility and performance.

Overview
Brand: BioTek™

Manufacturer Part Number: CYT5M

Cytation 5 Monochromator Fluorescence,Time-Resolved Fluores (2°mode), Luminescence,UV-Vis Absorbance

Code: NEW

Additional Details:
Additional Details: Weight: 36.30000kg



Product Code. 15340207

Quantity Price
1 £ 37800.0 / Each
Estimated Shipment date
from Supplier 20-03-2017
Add to basket


Description and Specification

Specification

Includes Gen5 software

  • Variable bandwidth monochromators, from 9nm to 50nm in 1nm increments for ultimate flexibility
  • Fluorescence, UV-Vis absorbance, luminescence, filtered luminescence, time resolved fluorescence, fluorescence polarization and laser-based Alpha detection modes available
  • Live cell assay support with CO2/O2 control, incubation to 65°C and shaking
  • Low volume (2μL) nucleic acid and protein quantification with the available Take3™ Plates
  • Dual reagent injector module available
  • Powerful yet easy to use Gen5 software for efficient plate reading, data analysis, exporting and reporting
  • Upgradable to add Fluorescence, brightfield, H&E and phase contrast microscopy and convention multi-mode detection provides powerful imaging and quantitative data collection in one instrument
Features
  • BTCYT5M: Monochromator fluorescence, monochromator UV-Vis absorbance, time resolved fluorescence (secondary mode) and luminescence; incubation to 65°C and linear, orbital and double-orbital shaking

Documentation