BioTek™ Cytation™ 5 Cell Imaging Multi-Mode Reader

For 6- to 384- well microplates, slides, Petri dishes and flasks. BioTek Cytation 5 Cell Imaging Multi-Mode Reader with Fluorescence, brightfield, H&E and phase contrast microscopy and upgradable multi-mode microplate reading provides powerful imaging and quantitative data collection in one instrument.

Overview
Brand: BioTek™

Manufacturer Part Number: CYT5MFV

Cytation 5 Monochromator Fluorescenc,Time-Resolved, Luminescence,UV-Vis, Top Filter Fluo,brightfield

Code: NEW

Additional Details:
Additional Details: Weight: 36.30000kg



Product Code. 15310217

Quantity Price
1 £ 77700.0 / Each
Estimated Shipment date
from Supplier 22-03-2017
Add to basket


Description and Specification

Specification

Includes One filter cube with two filter pairs (default blue - ex 360/40 nm, mirror 400 nm cutoff, em 460/40 nm and green - ex 485/20 nm, mirror 510 nm cutoff, em 528/20 nm); Gen5 standard software
For Use With (Application) Fluorescence, brightfield, H&E imaging, plus monochromator and filter-based multi-mode detection
Model CYT5MFV
Product Type Cell Imaging Multi-Mode Reader

  • 6 position objective turret; 2.5X to 60X magnification available
  • 4X, 10X, 20X and 40X phase contrast objectives available
  • Linear, orbital and double-orbital shaking
  • Live cell imaging and assay support with CO2/O2 control, incubation to 65°C and shaking optimize cell-based and other assays
  • Upgradable to add patented Hybrid Technology™ in multi-mode detection combines high performance filters with variable bandwidth monochromators for versatility and performance
  • The fluorescence monochromators offer a variable bandwidth from 9nm to 50nm in 1nm increments for ultimate flexibility
  • Laser-based excitation for AlphaScreen™ / AlphaLISA™ assays
  • CO2/O2 control, incubation to 65°C and shaking optimize cell-based and other assays
  • Low volume (2μL) nucleic acid and protein quantification with the available Take3™ Plates
  • Powerful yet easy to use Gen5 software for efficient plate reading, image capture and analysis
Features
  • BTCYT5MFV: Fluorescence, brightfield and H&E imaging with monochromator fluorescence, filter/dichroic fluorescence, monochromator UV-Vis absorbance, fluorescence polarization, time resolved fluorescence, luminescence, filtered luminescence incubation to 65C and linear, orbital and double-orbital shaking

Documentation