Learn More
BioTek™ Cytation™ 5 Cell Imaging Multi-Mode Reader
For 6- to 384- well microplates, slides, Petri dishes and flasks. BioTek Cytation 5 Cell Imaging Multi-Mode Reader with Fluorescence, brightfield, H&E and phase contrast microscopy and upgradable multi-mode microplate reading provides powerful imaging and quantitative data collection in one instrument.
Brand:  BioTek™ CYT5MPV
Code : NEW
Additional Details : Weight : 36.30000kg
 View more versions of this product
Product Code. 15300217
  • £74000.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • 6 position objective turret; 2.5X to 60X magnification available
  • 4X, 10X, 20X and 40X phase contrast objectives available
  • Linear, orbital and double-orbital shaking
  • Live cell imaging and assay support with CO2/O2 control, incubation to 65°C and shaking optimize cell-based and other assays
  • Upgradable to add patented Hybrid Technology™ in multi-mode detection combines high performance filters with variable bandwidth monochromators for versatility and performance
  • The fluorescence monochromators offer a variable bandwidth from 9nm to 50nm in 1nm increments for ultimate flexibility
  • Laser-based excitation for AlphaScreen™ / AlphaLISA™ assays
  • CO2/O2 control, incubation to 65°C and shaking optimize cell-based and other assays
  • Low volume (2μL) nucleic acid and protein quantification with the available Take3™ Plates
  • Powerful yet easy to use Gen5 software for efficient plate reading, image capture and analysis
Features
  • BTCYT5MPV: Fluorescence, brightfield, phase contrast and H&E imaging with monochromator fluorescence, monochromator UV-Vis absorbance, time resolved fluorescence (secondary mode), luminescence; incubation to 65C and linear, orbital and double-orbital shaking
Specifications
Specifications
Cell Imaging Multi-Mode Reader
One filter cube with two filter pairs (default blue - ex 360/40 nm, mirror 400 nm cutoff, em 460/40 nm and green - ex 485/20 nm, mirror 510 nm cutoff, em 528/20 nm); Gen5 standard software
Fluorescence, brightfield, H&E, phase contrast imaging and monochromator multi-mode detection
CYT5MPV
Videos
Documentation Special Offers
Special Offers