missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ Cytation™ 3 Multi-Mode Reader

Combines automated digital microscopy and conventional microplate detection. BioTek Cytation 3 Multi-Mode Reader with its unique design provides rich phenotypic cellular information with well-based quantitative data.

Brand:  BioTek™ CYT3MF

Code : NEW

Additional Details : Weight : 36.30000kg

 View more versions of this product

Product Code. 15391460

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Automated digital microscopy and/or multi-mode microplate detection in one instrument
  • Modular and upgradable: microscopy only, microplate reading only, or both
  • Flexible sample format: 6- to 384-well plates, microscope slides, cell culture flasks and Petri dishes
  • Affordable automation: automated XY stage, auto focus, auto exposure, automated image capture, LED intensity
  • Inverted fluorescence and brightfield microscopy
  • High Power LED cubes that provide superior excitation energy and long life
  • Optimized epifluorescence filter cubes for superior sensitivity for commonly used fluorophores: DAPI, CFP, GFP, YFP, RFP, Texas Red, CY5 and CY7
  • From 2.5x for full-well imaging to 60x for sub-micron resolution for intra-cellular details
  • Live cell assays with temperature control up to 45°C, CO2/O2 gas control accessory
  • Endpoint, time-lapse and montage assays
  • Automatic cell counting, sub-population analysis and image statistics
  • Dual reagent injectors
  • Patented Hybrid Technology™ microplate reading mode with high performance filter-based optics and high-flexibility quadruple monochromator optics
Note: Models BTCYT3MF. BTCYT3M, BTCYT3F can be upgraded to add fluorescence and brightfield imaging. Other models are available that include fluorescence and brightfield imaging; see Cat. Nos. BTCYT3V, BTCYT3FV, BTCYT3MV, BTCYT3MFV.
Specifications

Specifications

Monochromator, Fluorescence and Absorbance; Filter Based
50.8cm
41.91cm
44.5cm
Videos
Documentation Special Offers

Special Offers