Learn More
BioTek™ Cytation™ 3 Cell Imaging Multi-Mode Reader
Combines automated digital microscopy and conventional microplate detection. BioTek Cytation 3 Cell Imaging Multi-Mode Reader with its unique design provides rich phenotypic cellular information with well-based quantitative data.
Brand: BioTek™ CYT3V
Code : NEW
Additional Details : Weight : 36.30000kg
Description
- Automated digital microscopy and/or multi-mode microplate detection in one instrument
- Modular and upgradable: microscopy only, microplate reading only, or both
- Flexible sample format: 6- to 384-well plates, microscope slides, cell culture flasks and Petri dishes
- Affordable automation: automated XY stage, auto focus, auto exposure, automated image capture, LED intensity
- Inverted fluorescence and brightfield microscopy
- High Power LED cubes that provide superior excitation energy and long life
- Optimized epifluorescence filter cubes for superior sensitivity for commonly used fluorophores: DAPI, CFP, GFP, YFP, RFP, Texas Red, CY5 and CY7
- From 2.5x for full-well imaging to 60x for sub-micron resolution for intra-cellular details
- High quality images: Olympus objectives, 16-bit gray scale CCD camera, Semrock filters
- Live cell assays with temperature control up to 45°C, CO2/O2 gas control accessory
- Endpoint, time-lapse and montage assays
- Automatic cell counting, sub-population analysis and image statistics
- Dual reagent injectors
- Patented Hybrid Technology™ microplate reading mode with high performance filter-based optics and high-flexibility quadruple monochromator optics
Specifications
Automated Fluorescence and Brightfield Imaging |
Technical Documents
Special Offers
-
Biotek Instruments
Biotek Cell Imaging and Multi-Mode Readers
Cytationâ„¢ 1 Cell Imaging Multi-Mode Reader eliminates the complexities of multi-mode detection without compromising performance.