Learn More
BioTek™ Cytation™ 3 Cell Imaging Multi-Mode Reader
Combines automated digital microscopy and conventional microplate detection. BioTek Cytation 3 Cell Imaging Multi-Mode Reader with its unique design provides rich phenotypic cellular information with well-based quantitative data.
Brand:  BioTek™ CYT3MV
Code : NEW
Additional Details : Weight : 36.30000kg
 View more versions of this product
Product Code. 15370217
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Automated digital microscopy and/or multi-mode microplate detection in one instrument
  • Modular and upgradable: microscopy only, microplate reading only, or both
  • Flexible sample format: 6- to 384-well plates, microscope slides, cell culture flasks and Petri dishes
  • Affordable automation: automated XY stage, auto focus, auto exposure, automated image capture, LED intensity
  • Inverted fluorescence and brightfield microscopy
  • High Power LED cubes that provide superior excitation energy and long life
  • Optimized epifluorescence filter cubes for superior sensitivity for commonly used fluorophores: DAPI, CFP, GFP, YFP, RFP, Texas Red, CY5 and CY7
  • From 2.5x for full-well imaging to 60x for sub-micron resolution for intra-cellular details
  • High quality images: Olympus objectives, 16-bit gray scale CCD camera, Semrock filters
  • Live cell assays with temperature control up to 45°C, CO2/O2 gas control accessory
  • Endpoint, time-lapse and montage assays
  • Automatic cell counting, sub-population analysis and image statistics
  • Dual reagent injectors
  • Patented Hybrid Technology™ microplate reading mode with high performance filter-based optics and high-flexibility quadruple monochromator optics
Specifications
Specifications
Monochromator; Automated Imaging
Videos
Documentation Special Offers
Special Offers