missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Buffer Bottle Assembly for 50 TS Microplate Washer

Accessory for 50 TS microplate washer

Brand:  Agilent BioTek 4070515

Code : 03

Additional Details : Weight : 0.01000kg

Product Code. 10433133

  • £103.00 / Each
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • For use with 50 TS Microplate Strip Washer
  • Includes: Bottle, cap w/fitting and tubing

Specifications

Specifications

2 L
67.62 oz.
ELx 405 Microplate Washer, Agilent BioTek Dispenser, Pipetting Systems with Non-autoclavable Fluid Paths
Buffer Bottle Assembly
Bottle, connector, cap with fitting and tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers