Learn More
Agilent BioTek BioStack Automated Microplate Stacker - Microplate Stacks
Storage stacks
Brand:  Agilent BioTek 7310023
Additional Details : Weight : 5.89670kg
 View more versions of this product
Product Code. 11563635
  • £3557.62 / Each
Estimated Shipment: 23-04-2024
to see stock.
Description
Description
  • One set
Specifications
Specifications
Microplates
Storage
100 Microplates
Bio-tek Biostack automated microplate stacker, unlidded plates ≤14 mm
2 stacks of 50 microplates