Learn More
Agilent BioTek Liquid Dispenser Accessory, Syringe Pump Dispense Tubing and Check Valve Kit
Provides compatiblity with DMSO and acetonitrile
Brand: Agilent BioTek 7180541
Additional Details : Weight : 1.36078kg
Description
Spare syringe pump dispense tubing and check valve kit: Teflon-lined, DMSO and acetonitrile compatible (non-autoclavable, but compatible with the autoclavable syringe module hardware).Specifications
Kit | |
EL406 Microplate Washer Dispenser or MultiFlo Dispenser | |
Tubing and check valve kit |