missing translation for 'onlineSavingsMsg'
Learn More

405™ Touch Microplate Washer

Configured with many options to meet myriad assay requirements

Brand:  Biotek Instruments 405TSHT

Code : Z1

Additional Details : Weight : 13.50000kg

 View more versions of this product

Product Code. 12553567

  • £19700.00 / Each


Estimated Shipment date
from Supplier 04-04-2018
Add to basket

Specifications

Specifications

1 to 250
Solid bottom plates: 3 asp./disp/ cycles, 300 uL/well, 96 wells, 96 tube manifold, final aspirate: <30s; 3 asp./disp/ cycles, 100 uL/well, 384 wells, 96 tube manifold, final aspirate: <80s; 1 asp./disp/ cycles, 40 uL/well, 384 wells, 192 tube manifold
2
35.6cm
25.4cm
50/60Hz
100 to 240V
ELISA, MSD assays, HCS immune cytochemistry, FLIPR Ca2+ flux, Cell-based assays, Magnetic and polystyrene bead processing (gene expression assays, cytokine assays), ELISPOT assays
ELISA, Cell-based assays (model dependent), Magnetic and polystyrene bead, Multiplex assays, Bead-based ELISAs, Filtration-to-waste processes
Internal positive displacement pump
100 to 240V 50/60Hz
Programmable in minutes and seconds, up to 60 min.
LED
High res LED backlit touch screen for instrument control
43.2cm
With Internal Buffer Switching: 16.5kg; Without Internal Buffer Switching: 13.5kg
Microplate Washer w/Ultrasonic advantage
≤2μL/well
25 to 3000μL/well
1 to 11
384
Videos