Learn More
BioTek™ 405™ Touch Microplate Washer with Verify™ Technology
Features patented Verify™ technology which automatically checks for manifold tube blockage and visually reports any failed wells. Ultrasonic Advantage™ can be used to automatically and thoroughly clean the aspirate and dispense manifold.
Brand:  BioTek™ 405TSRSQ
Additional Details : Weight : 13.50000kg
 View more versions of this product
Product Code. 15191259
  • £14499.00 / Each
Estimated Shipment date
from Supplier 20-03-2018
Add to basket
Description
Description
Hardware features and options:
  • Patented built-in Verify sensor automatically checks dispense / aspirate performance and reports clogged locations in the color graphical display.
  • Accommodates 96- and 384-well microplates
  • High resolution touch screen user interface
  • Onboard computer running Microsoft Windows CE
  • Patent pending built-in Ultrasonic Advantage automatically cleans clogged tubes
  • Two USB flash drive ports enable convenient file transfer, storage and operation
  • Automated internal 4-buffer switching
  • Internal positive displacement pump eliminates use of troublesome pressure delivery systems and allows use of any size dispense reservoir
  • Built-in priming trough allows automatic manifold priming and rinsing during both operation and maintenance modes and eliminates need for blank plates when used in automated robotic systems
  • All washers vacuum filtration ready (accessory filtration modules sold separately)
  • All washers biomagnetic separation ready (accessory magnets sold separately)
405 Select TS:
  • 96- or 384-well washing with patented 96-tube Dual-Action™ manifold
  • Cell washing is optimized with two low-flow rates and 20° angled dispense tubes ideal for washing loosely adherent cell monolayers
405 HT TS:
  • Fast 384-well washing with patented 192-tube Dual-Action™ manifold for independent dispense and aspiration control for precise overfill washing and overflow protection
  • Superior tube-in-tube aspiration manifold design for 384-well washing with buffers not containing surfactant
  • 96-well washing ready with optional 96-tube Dual-Action manifold
405 TS:
  • 405 TS models automate basic 96-well dispensing and aspiration
Software Features:
  • Context sensitive help system complete with recommendations, diagrams and images is only a key press away when questions arise
  • Pre-defined protocols and instructional videos walk users through instrument setup and basic use
  • Fully customizable protocol parameters for individualized wash requirements - gentle cell-washing to vigorous wash for high-binding assays
  • AutoPrime
  • Additional software flexibility and 21 CFR Part 11 compliance with Liquid Handling Control™ (LHC) Software for method development, downloading and washing plates.
  • Run protocols created onboard, downloaded from LHC™ Software or transferred on USB drive
Specifications
Specifications
Microplate Washer
96
100 to 240VAC, 50/60Hz, ≤8.0 Ampere
Ultrasonic Advantage™, Verify™ technology, Biomagnetic separation, Vacuum filtration
Videos