missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ 405™ Select LS Microplate Washers for 96/384 Well Microplate Washing

Fifth generation washer automates basic 96- or 384-well dispensing and aspiration

Brand:  BioTek™ 405LSU

Additional Details : Weight : 13.50000kg

 View more versions of this product

Product Code. 12513567

  • £15180.00 / Each

Estimated Shipment
09-04-2021

Please to purchase this item. Need a web account? Register with us today!


Requires : Required for operation: BT1170530, Complete Dispense/Waste System, 4L bottles. Must order separately.

Description

Description

  • For easy processing of cell based assays, ELISA and multiplex assays
  • 96/384 well washing with patented 96 tube Dual Action™ manifold
  • Angled dispense tubes, extensive aspirate and dispense rates, plate position controls provide optimal conditions for washing even fragile cell layers
  • Cell washing optimized with two low-flow rates and 20° angled dispense tubes ideal for washing loosely adherent cell monolayers
  • Programmable in minutes and seconds, up to 60 minutes (variable intensities)
  • Waste and vacuum sensing, fluid and flow detection
  • Aerosol cover and overflow protection
Hardware
  • Accommodates 96 or 384 well microplates
  • 2 x 24 character backlit LCD display and membrane keypad user interface
  • Automated internal four buffer switching
  • Internal positive displacement pump eliminates use of troublesome pressure delivery systems and allows use of any size dispense reservoir
  • Built-in priming trough allows automatic manifold priming and rinsing during both operation and maintenance modes
  • Quick change manifold design
  • BioStack and third party automation compatible
  • All washers biomagnetic separation ready (accessory magnets sold separately)
  • All washers vacuum filtration ready (accessory filtration modules sold separately)
Software
Quick menu allows fluid paths to be easily primed, wash process started and manifold cleaned
Predefined protocols included for out-of-box washing
Fully customizable protocol parameters for individualized wash requirements (gentle cell washing to vigorous wash for high binding assays)
AutoPrime and auto maintenance routines
  • Run protocols created onboard or downloaded from LHC Software
  • Order Info

    Required for operation: BT1170530, Complete Dispense/Waste System, 4L bottles. Must order separately.

    Specifications

    Specifications

    50/60Hz
    100 to 240V AC
    15° to 30°C
    100 to 240V AC 50/60 Hz ≤8A
    Washer for 96- or 384-well microplate
    10 to 85%, non condensing
    96 or 384
    Videos
    SDS
    Documents