Learn More
BioTek™ 405™ LS Microplate Washer with Verify™ Technology
Brand:  BioTek™ 405LSRSQ
Additional Details : Weight : 13.50000kg
 View more versions of this product
Product Code. 15396186
  • £12500.00 / Each
Estimated Shipment date
from Supplier 22-02-2018
Add to basket
Description
Description
405 LS washer with Ultrasonic Advantage™ and Verify™ technology
Specifications
Specifications
100 - 240V
25 to 3,000μL/well
100 - 240 Volts AC 50/60 Hz, <8.0 A
96-well
Videos