Learn More
BioTek™ 405™ LS Microplate Washer
Accurate, precise and fast washing
Brand:  BioTek™ 405LSHT
Code : Z1
Additional Details : Weight : 13.50000kg
Product Code. 12553557
  • £18100.00 / Each
Estimated Shipment date
from Supplier 02-07-2018
Add to basket
Description
Description
  • For 384-well microplates
  • Ultrasonic Advantage™
Specifications
Specifications
1 to 250
16.5kg
100 to 240V
15° to 30°C
Programmable in min. and sec., up to 60 min.
Ultrasonic Advantage
CE, TUV
100 to 240VAC, 50/60Hz, <8A
Videos