Learn More
Teknon™ Biocleanse Concentrate
Use in scientific, medical and biotechnology environments to combat biological hazards. Teknon™ Biocleanse Concentrate is a concentrated deodorising cleaner, degreaser, disinfectant and fungicide.
Brand: Teknon™ D/0006/21
Code : 04
Additional Details : CAS Number : 68424-85-1 Weight : 5.25500kg Transport : UN number : 1903 Chem class : 8 Pack group : II
Description
This product has powerful bactericidal and virucidal properties. Biocleanse concentrate is free from sodium hypochlorite, phosphates and enzymes and effectively removes blood, fat, proteins, grease and other organic and non-organic contaminants whilst simultaneously disinfecting the treated surface. It is safe to use on ferrous and non-ferrous metals, ceramics, glassware and plastics.
Bioclease concentrate exceeds the requirements of BS EN 1276, killing E. coli, Salmonella, Listeria, Candida and Penicillium at 1% dilution and MRSA at 2% dilution. Efficacy against C. diff, Avian Flu, HIV and HBV is proven at 5% dilution.
- Under COSHH Regulations, does not require maximum exposure limit
- When diluted to normal use concentrations, is not irritating to skin and non-tainting
- Wide spectrum biocide can be used with ultrasonic cleaning equipment
- Density: 1.051 at 20°C
- Vapor Pressure: 17.5mm Hg at 20°C
Specifications
Concentrated Liquid | |
Blue | |
Cannister | |
For use in scientific, medical and biotechnology environments to combat biological hazards |
10 (1%, aq. soln.) | |
5–15% alkaline sequestrant, 1–5% nonionic surfactant, 5–15% benzalkonium chloride | |
Store in a cool, dry place protected from frost and away from acids and strong oxidising agents. Store upright in original containers | |
5 L |
Safety and Handling
- BIOCLEANSE
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : lactic acid