Learn More
Thermo Scientific™ BioBasic™ SEC 120 Size Exclusion Chromatography HPLC Columns
Outstanding size separation of water soluble compounds.
Brand: Thermo Scientific™ 73405-037821
Code : 16
Additional Details : Weight : 0.10000kg
Description
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 120 Size Exclusion Chromatography HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straightforward method development, simple mobile phases
- 120Å pore size
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
Guard Column | |
Size Exclusion Chromatography | |
5800 psi (400 bar) | |
120 Å | |
Diol (OH) | |
Silica | |
Size Exclusion |
7.8 mm | |
30 mm | |
5 μm | |
L33 | |
BioBasic | |
Size Exclusion | |
Guard Column |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.