missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ SEC 60 Size Exclusion Chromatography HPLC Columns

Outstanding size separation of water soluble compounds.

Brand:  Thermo Scientific™ 73305-037821

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10669295

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 60 Size Exclusion Chromatography HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 60Å pore size
  • Particle size: 5μm

Ordering Information:

For information on bulk quantities, please inquire.

Specifications

Specifications

Guard Column
Size Exclusion Chromatography
5800 psi (400 bar)
60 Å
Diol (OH)
Base-Deactivated Silica
Size Exclusion
7.8 mm
30 mm
5 μm
L33
BioBasic
Size Exclusion
Guard Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers