missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ AX LC Columns

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

Brand:  Thermo Scientific™ 73105-103030

UNSPSC : 41115709

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10477244

  • £449.00 / Each


Estimated Shipment
22-08-2017
Add to basket

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


Specifications

Description

  • 5μm particle size, 300Å pore size
  • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
  • Low hydrophobicity
  • Capacity of 0.22mEq/g; 3% carbon load
  • Use for normal phase and hydrophilic interaction chromatography (HILIC)
  • Stable under demanding pH conditions (2 to 8 pH range)
Description & Specifications

Specifications

3%
Ion Exchange
Yes
100mm
Silica, Spherical Fully Porous Ultrapure
2 to 8
100m²/g
L7
Polyethyleneimine
Analytical Column
3mm
Low
400bar
5μm
300Å
60°C
Anion Exchange