missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BioBasic™ C4 Reversed Phase HPLC Column, 5 μm, 4 mm x 150 mm
Column Format: Analytical Column; Carbon Load: 4%; Pore Size: 300 Å; Column Phase: Reversed Phase
Brand: Thermo Scientific™ 72305-154030
Code : 16
Additional Details : Weight : 0.10000kg
Description
- Based on 300Å silica for outstanding biomolecule performance
- Lower carbon load for optimal retention of larger peptides an proteins
- Outstanding reproducibility, efficiency and column lifetime
- Excellent for LC/MS separations
Specifications
Analytical Column | |
Low | |
5 μm | |
300 Å | |
L26 | |
Silica | |
Reversed Phase | |
Reversed Phase |
4 mm | |
150 mm | |
2 to 9 | |
100 m²/g | |
C4 | |
4% | |
Yes |