missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ 4 LC Columns

BioBasic 4 LC Columns are designed for the analysis of larger proteins and peptides.

Brand:  Thermo Scientific™ 72305-011001

UNSPSC : 41115709

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10554685

  • £243.00 / Pack of 4


Estimated Shipment
23-08-2017
Add to basket

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


Specifications

Description

  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.

Description & Specifications

Specifications

4%
Reversed Phase
Yes
400bar
2 to 9
100m²/g
Guard Cartridge, 4 PK
C4
Guard Cartridge, 4 PK
1mm
10mm
5μm
300Å
60°C
L26