missing translation for 'onlineSavingsMsg'
Learn More

BIO-acetoxime, Tocris Bioscience™

Selective GSK-3α/β inhibitor

Brand:  Tocris (Bio-Techne) 387410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13443744

  • £230.79 / 10mg

Estimated Shipment date
from Supplier 28-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective GSK-3α/β inhibitor (IC50 values are 0.01, 2.4, 4.3 and 63 μM for GSK-3α/β, CDK5/p25, CDK2/cyclin A and CDK1/cyclin B respectively). Induces Wnt signaling and inhibits CD8+ T cell effector differentiation.
Specifications

Chemical Identifiers

667463-85-6
398.216
bio-acetoxime, gsk-3 inhibitor x, bio-acetoxime gsk-3 inhibitor x, e,3e-6'-bromo-2'-oxo-1h,1'h-2,3'-biindolyliden-3-ylidene amino acetate
[[(2E)-2-(6-bromo-2-oxo-1H-indol-3-ylidene)indol-3-yl]amino] acetate
C18H12BrN3O3
ORWSYHWTBSHFIV-BMRADRMJSA-N
77519830
CC(=O)ONC1=C2C=CC=CC2=NC1=C3C4=C(C=C(C=C4)Br)NC3=O

Specifications

10mg
667463-85-6
bio-acetoxime, gsk-3 inhibitor x, bio-acetoxime gsk-3 inhibitor x, e,3e-6'-bromo-2'-oxo-1h,1'h-2,3'-biindolyliden-3-ylidene amino acetate
CC(=O)ONC1=C2C=CC=CC2=NC1=C3C4=C(C=C(C=C4)Br)NC3=O
398.216
398.21
BIO-acetoxime
C18H12BrN3O3
ORWSYHWTBSHFIV-BMRADRMJSA-N
[[(2E)-2-(6-bromo-2-oxo-1H-indol-3-ylidene)indol-3-yl]amino] acetate
77519830
>98%
Videos
Documents