missing translation for 'onlineSavingsMsg'
Learn More

BIMU 8, Tocris Bioscience™

Potent 5-HT4 receptor full agonist

Brand:  RD Systems (Bio-Techne) 4374/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13464644

  • £620.77 / 50mg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

Potent 5-HT4 receptor full agonist (KD = 77 nM and EC50 = 18 nM for wild type 5-HT4 receptors). Binds to receptors in a mechanism different to that of the endogenous ligand 5-HT.
Specifications

Chemical Identifiers

134296-40-5
378.901
bimu 8, bimu8, bimu-8, endo-n-8-methyl-8-azabicyclo-3.2.1 oct-3-yl-2,3-dihydro-3-isopropyl-2-oxo-1h-benzimidazol-1-carboxamide, 1h-benzimidazole-1-carboxamide, 2,3-dihydro-n-8-methyl-8-azabicyclo 3.2.1 oct-3-yl-3-1-methylethyl-2-oxo-, monohydrochloride, endo, 2,3-dihydro-n-3-endo-8-methyl-8-azabicyclo 3.2.1 oct-3-yl-3-1-methylethyl-2-oxo-1h-benzimidazole-1-carboxamide hydrochloride, n-1r,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl-2-oxo-3-propan-2-ylbenzimidazole-1-carboxamide hydrochloride
N-[(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl]-2-oxo-3-propan-2-ylbenzimidazole-1-carboxamide;hydrochloride
C19H27ClN4O2
NQYXXIUVFVOJCX-XZPOUAKSSA-N
5311028
CC(C)N1C2=CC=CC=C2N(C1=O)C(=O)NC3CC4CCC(C3)N4C.Cl

Specifications

BIMU 8
134296-40-5
bimu 8, bimu8, bimu-8, endo-n-8-methyl-8-azabicyclo-3.2.1 oct-3-yl-2,3-dihydro-3-isopropyl-2-oxo-1h-benzimidazol-1-carboxamide, 1h-benzimidazole-1-carboxamide, 2,3-dihydro-n-8-methyl-8-azabicyclo 3.2.1 oct-3-yl-3-1-methylethyl-2-oxo-, monohydrochloride, endo, 2,3-dihydro-n-3-endo-8-methyl-8-azabicyclo 3.2.1 oct-3-yl-3-1-methylethyl-2-oxo-1h-benzimidazole-1-carboxamide hydrochloride, n-1r,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl-2-oxo-3-propan-2-ylbenzimidazole-1-carboxamide hydrochloride
CC(C)N1C2=CC=CC=C2N(C1=O)C(=O)NC3CC4CCC(C3)N4C.Cl
378.901
378.9
50mg
C19H27ClN4O2
NQYXXIUVFVOJCX-XZPOUAKSSA-N
N-[(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl]-2-oxo-3-propan-2-ylbenzimidazole-1-carboxamide;hydrochloride
5311028
>98%
Videos