missing translation for 'onlineSavingsMsg'
Learn More

Bilirubin, 99%, ACROS Organics™

Brand:  Acros Organics 230220050

Code : 29

Additional Details : CAS Number : 635-65-4 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10559823

  • £241.40 / 5g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Bilirubin
98.5
99%
584.67
635-65-4
100.0
C33H36N4O6
MFCD00005499

Specifications

Bilirubin
98.5
Authentic
59600L/(mol.cm) min. at lambda max 452nm +/- 2nm in CHCl3
98.5% min.
99%
584.67
Orange to reddish-brown
5g
635-65-4
100.0
1.8% max. (60°C, 2 hrs) (vacuum)
Glass bottle
Fine Crystalline Powder
C33H36N4O6
MFCD00005499
15, 1222
Videos
SDS