Learn More
Bifemelane hydrochloride, Tocris Bioscience™
MAO-A and MAO-B inhibitor
Brand:  Tocris (Bio-Techne) 076750
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13487973
  • £461.52 / 50mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Antidepressant MAO inhibitor. Reverses catalepsy induced by tetrabenazine in mice and increases locomotor activity in MPTP-treated marmosets.
Specifications
Chemical Identifiers
62232-46-6
305.846
4-(2-Benzylphenoxy)-N-methylbutylamine hydrochloride
C18H24ClNO
MEAHDXWXNNDSAK-UHFFFAOYSA-N
6917789
Specifications
Bifemelane hydrochloride
>99%
4-(2-Benzylphenoxy)-N-methylbutylamine hydrochloride
MEAHDXWXNNDSAK-UHFFFAOYSA-N
4-(2-benzylphenoxy)-N-methylbutan-1-amine;hydrochloride
6917789
62232-46-6
C18H24ClNO
50mg
CNCCCCOC1=CC=CC=C1CC2=CC=CC=C2.Cl
305.846
305.84
Videos