missing translation for 'onlineSavingsMsg'
Learn More

(-)-Bicuculline methochloride, Tocris Bioscience™

GABAA antagonist; more water soluble version of (+)-bicuculline (Cat. No. 0130)

Brand:  Tocris (Bio-Techne) 0131/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13467443

  • £127.89 / 10mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Methochloride salt of (+)-bicuculline . Water soluble and more stable than bicuculline. Non-GABA receptor-mediated actions reported.

Methoiodide Salt and Methobromide Salt also available.

Specifications

Chemical Identifiers

53552-05-9
417.842
--bicuculline methochloride
(6R)-6-[(5R)-6,6-dimethyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium-5-yl]-6H-furo[3,4-g][1,3]benzodioxol-8-one;chloride
C21H20ClNO6
RLJKFAMYSYWMND-STYNFMPRSA-M
124080984
C[N+]1(CCC2=CC3=C(C=C2C1C4C5=C(C6=C(C=C5)OCO6)C(=O)O4)OCO3)C.[Cl-]

Specifications

(-)-Bicuculline methochloride
53552-05-9
--bicuculline methochloride
C[N+]1(CCC2=CC3=C(C=C2C1C4C5=C(C6=C(C=C5)OCO6)C(=O)O4)OCO3)C.[Cl-]
417.842
417.85
10mg
C21H20ClNO6
RLJKFAMYSYWMND-STYNFMPRSA-M
(6R)-6-[(5R)-6,6-dimethyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium-5-yl]-6H-furo[3,4-g][1,3]benzodioxol-8-one;chloride
124080984
>99%
Videos
Documents